Assay Details
| Gene Symbol: Fhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869223.2 | XP_004869280.1 | 7 - 8 | 1124 | 54 | |
| XM_021247868.1 | XP_021103527.1 | 7 - 8 | 1284 | 54 | ||
More Information
Set Membership: |
|

Fhdc1
Genomic Map