Assay Details
| Gene Symbol: LOC101699218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870619.2 | XP_004870676.1 | 2 - 3 | 258 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101699218| Gene Symbol: LOC101699218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870619.2 | XP_004870676.1 | 2 - 3 | 258 | 109 | |
Set Membership: |
|