Assay Details
| Gene Symbol: Sh2d6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072183.2 | XP_012927637.1 | 8 - 9 | 4480 | 61 | |
| XM_021258213.1 | XP_021113872.1 | 6 - 7 | 722 | 61 | ||
| XM_021258214.1 | XP_021113873.1 | 7 - 8 | 757 | 61 | ||
| XM_021258215.1 | XP_021113874.1 | 7 - 8 | 549 | 61 | ||
| XM_021258216.1 | XP_021113875.1 | 5 - 6 | 847 | 61 | ||
| XM_021258217.1 | XP_021113876.1 | 7 - 8 | 666 | 61 | ||
| XM_021258218.1 | XP_021113877.1 | 5 - 6 | 518 | 61 | ||
More Information
Set Membership: |
|

Sh2d6
Genomic Map