Assay Details
| Gene Symbol: Spic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072246.2 | XP_012927700.1 | 3 - 4 | 198 | 60 | |
| XM_021258420.1 | XP_021114079.1 | 3 - 4 | 217 | 60 | ||
More Information
Set Membership: |
|

Spic
Genomic Map