Assay Details
Gene Symbol: Icos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013074480.2 | XP_012929934.1 | 3 - 4 | 1049 | 95 | |
XM_021263577.1 | XP_021119236.1 | 5 - 6 | 1113 | 95 | ||
XM_021263578.1 | XP_021119237.1 | 4 - 5 | 796 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
