Assay Details
| Gene Symbol: LOC101702367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021240035.1 | XP_021095694.1 | 21 - 22 | 2962 | 98 | |
| XM_021240036.1 | XP_021095695.1 | 21 - 22 | 2962 | 98 | ||
| XM_021240037.1 | XP_021095696.1 | 21 - 22 | 2962 | 98 | ||
| XM_021240038.1 | XP_021095697.1 | 21 - 22 | 2963 | 98 | ||
More Information
Set Membership: |
|

LOC101702367
Genomic Map