Assay Details
| Gene Symbol: LOC101701112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246390.1 | XP_021102049.1 | 3 - 4 | 912 | 98 | |
| XM_021246391.1 | XP_021102050.1 | 3 - 4 | 911 | 98 | ||
| XM_021246392.1 | XP_021102051.1 | 4 - 5 | 547 | 98 | ||
| XM_021246393.1 | XP_021102052.1 | 2 - 3 | 269 | 98 | ||
| XM_021246394.1 | XP_021102053.1 | 3 - 4 | 2149 | 98 | ||
More Information
Set Membership: |
|

LOC101701112
Genomic Map