Assay Details
| Gene Symbol: LOC101701112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246390.1 | XP_021102049.1 | 8 - 9 | 1565 | 73 | |
| XM_021246391.1 | XP_021102050.1 | 8 - 9 | 1564 | 73 | ||
| XM_021246392.1 | XP_021102051.1 | 9 - 10 | 1200 | 73 | ||
| XM_021246393.1 | XP_021102052.1 | 7 - 8 | 922 | 73 | ||
| XM_021246394.1 | XP_021102053.1 | 8 - 9 | 2802 | 73 | ||
More Information
Set Membership: |
|

LOC101701112
Genomic Map