Assay Details
| Gene Symbol: LOC101701112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246390.1 | XP_021102049.1 | 9 - 10 | 1756 | 70 | |
| XM_021246392.1 | XP_021102051.1 | 10 - 11 | 1391 | 70 | ||
| XM_021246393.1 | XP_021102052.1 | 8 - 9 | 1113 | 70 | ||
More Information
Set Membership: |
|

LOC101701112
Genomic Map