Assay Details
| Gene Symbol: Ppp2r5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837423.2 | XP_004837480.1 | 13 - 14 | 1919 | 70 | |
| XM_004837424.3 | XP_004837481.1 | 13 - 14 | 1922 | 70 | ||
| XM_021247820.1 | XP_021103479.1 | 12 - 13 | 1390 | 70 | ||
| XM_021247821.1 | XP_021103480.1 | 13 - 14 | 1387 | 70 | ||
More Information
Set Membership: |
|

Ppp2r5e
Genomic Map