Assay Details
| Gene Symbol: Jph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842042.2 | XP_004842099.1 | 3 - 4 | 1495 | 73 | |
| XM_004842043.2 | XP_004842100.1 | 3 - 4 | 1495 | 73 | ||
| XM_013071217.2 | XP_012926671.1 | 3 - 4 | 1495 | 73 | ||
More Information
Set Membership: |
|

Jph1
Genomic Map