Assay Details
| Gene Symbol: Ripor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843027.3 | XP_004843084.1 | 6 - 7 | 551 | 81 | |
| XM_004843029.3 | XP_004843086.1 | 6 - 7 | 548 | 81 | ||
| XM_013071548.2 | XP_012927002.1 | 6 - 7 | 552 | 81 | ||
| XM_013071549.2 | XP_012927003.1 | 6 - 7 | 552 | 81 | ||
| XM_021256843.1 | XP_021112502.1 | 5 - 6 | 527 | 81 | ||
| XM_021256844.1 | XP_021112503.1 | 6 - 7 | 620 | 81 | ||
| XM_021256845.1 | XP_021112504.1 | 3 - 4 | 156 | 81 | ||
| XM_021256846.1 | XP_021112505.1 | 6 - 7 | 552 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021256845.1 transcripts.
Set Membership: |
|

Ripor1
Genomic Map