Assay Details
| Gene Symbol: Tbc1d7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847677.3 | XP_004847734.1 | 7 - 8 | 910 | 85 | |
| XM_004847678.3 | XP_004847735.1 | 7 - 8 | 905 | 85 | ||
| XM_004847679.3 | XP_004847736.1 | 6 - 7 | 829 | 85 | ||
| XM_021260641.1 | XP_021116300.1 | 6 - 7 | 790 | 85 | ||
| XM_021260642.1 | XP_021116301.1 | 7 - 8 | 1009 | 85 | ||
| XM_021260643.1 | XP_021116302.1 | 6 - 7 | 935 | 85 | ||
| XM_021260644.1 | XP_021116303.1 | 6 - 7 | 826 | 85 | ||
More Information
Set Membership: |
|

Tbc1d7
Genomic Map