Assay Details
| Gene Symbol: Fopnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855785.2 | XP_004855842.1 | 4 - 5 | 590 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fopnl| Gene Symbol: Fopnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855785.2 | XP_004855842.1 | 4 - 5 | 590 | 78 | |