Assay Details
| Gene Symbol: Arf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863056.3 | XP_004863113.1 | 4 - 5 | 662 | 63 | |
| XM_021243169.1 | XP_021098828.1 | 4 - 5 | 612 | 63 | ||
More Information
Set Membership: |
|

Arf3
Genomic Map