Assay Details
| Gene Symbol: Slc25a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865626.3 | XP_004865683.1 | 5 - 6 | 808 | 56 | |
| XM_021245132.1 | XP_021100791.1 | 5 - 6 | 820 | 56 | ||
| XM_021245133.1 | XP_021100792.1 | 5 - 6 | 822 | 56 | ||
More Information
Set Membership: |
|

Slc25a23
Genomic Map