Assay Details
| Gene Symbol: Cnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865983.3 | XP_004866040.1 | 4 - 5 | 493 | 87 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnn2| Gene Symbol: Cnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865983.3 | XP_004866040.1 | 4 - 5 | 493 | 87 | |