Assay Details
| Gene Symbol: Taar5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921230.2 | XP_004921287.2 | 1 - 2 | 101 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004921230.2 transcripts.
Set Membership: |
|

Taar5
Genomic Map