Assay Details
Gene Symbol: LOC101712837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245921.1 | XP_021101580.1 | 3 - 4 | 590 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021245921.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
