Assay Details
Gene Symbol: LOC101712837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245921.1 | XP_021101580.1 | 7 - 8 | 1384 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
