Assay Details
| Gene Symbol: Zdhhc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838304.2 | XP_004838361.1 | 3 - 4 | 784 | 75 | |
| XM_013069668.2 | XP_012925122.1 | 3 - 4 | 620 | 75 | ||
| XM_021252864.1 | XP_021108523.1 | 3 - 4 | 355 | 75 | ||
| XM_021252865.1 | XP_021108524.1 | 4 - 5 | 947 | 75 | ||
More Information
Set Membership: |
|

Zdhhc21
Genomic Map