Assay Details
| Gene Symbol: LOC101699770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841315.3 | XP_004841372.1 | 11 - 12 | 1887 | 99 | |
| XM_021255417.1 | XP_021111076.1 | 10 - 11 | 1822 | 99 | ||
More Information
Set Membership: |
|

LOC101699770
Genomic Map