Assay Details
| Gene Symbol: LOC101699770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841315.3 | XP_004841372.1 | 8 - 9 | 1377 | 115 | |
| XM_021255417.1 | XP_021111076.1 | 7 - 8 | 1312 | 115 | ||
More Information
Set Membership: |
|

LOC101699770
Genomic Map