Assay Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841697.3 | XP_004841754.1 | 2 - 3 | 636 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004841697.3 transcripts.
Set Membership: |
|

Rfesd
Genomic Map