Assay Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841697.3 | XP_004841754.1 | 4 - 5 | 845 | 72 | |
| XM_004841698.3 | XP_004841755.1 | 3 - 4 | 749 | 72 | ||
| XM_021255785.1 | XP_021111444.1 | 3 - 4 | 823 | 72 | ||
| XM_021255786.1 | XP_021111445.1 | 3 - 4 | 823 | 72 | ||
More Information
Set Membership: |
|

Rfesd
Genomic Map