Assay Details
| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844523.3 | XP_004844580.1 | 9 - 10 | 2274 | 60 | |
| XM_013072169.2 | XP_012927623.1 | 8 - 9 | 1696 | 60 | ||
| XM_013072170.2 | XP_012927624.1 | 8 - 9 | 1740 | 60 | ||
More Information
Set Membership: |
|

Slc20a1
Genomic Map