Assay Details
| Gene Symbol: LOC101699327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847888.1 | XP_004847945.1 | 2 - 3 | 274 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101699327| Gene Symbol: LOC101699327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847888.1 | XP_004847945.1 | 2 - 3 | 274 | 84 | |
Set Membership: |
|