Assay Details
Gene Symbol: Bmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847995.3 | XP_004848052.1 | 1 - 2 | 350 | 111 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
