Assay Details
| Gene Symbol: Virma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850114.3 | XP_004850171.1 | 14 - 15 | 3858 | 106 | |
| XM_021262644.1 | XP_021118303.1 | 14 - 15 | 3860 | 106 | ||
| XM_021262645.1 | XP_021118304.1 | 13 - 14 | 3534 | 106 | ||
| XM_021262646.1 | XP_021118305.1 | 11 - 12 | 3359 | 106 | ||
| XM_021262647.1 | XP_021118306.1 | 14 - 15 | 3863 | 106 | ||
| XM_021262648.1 | XP_021118307.1 | 8 - 9 | 2860 | 106 | ||
More Information
Set Membership: |
|

Virma
Genomic Map