Assay Details
| Gene Symbol: Virma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850114.3 | XP_004850171.1 | 20 - 21 | 4876 | 90 | |
| XM_021262644.1 | XP_021118303.1 | 20 - 21 | 4878 | 90 | ||
| XM_021262645.1 | XP_021118304.1 | 19 - 20 | 4552 | 90 | ||
| XM_021262646.1 | XP_021118305.1 | 17 - 18 | 4377 | 90 | ||
| XM_021262647.1 | XP_021118306.1 | 20 - 21 | 4881 | 90 | ||
| XM_021262648.1 | XP_021118307.1 | 14 - 15 | 3878 | 90 | ||
More Information
Set Membership: |
|

Virma
Genomic Map