Assay Details
| Gene Symbol: Virma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850114.3 | XP_004850171.1 | 5 - 6 | 714 | 73 | |
| XM_021262644.1 | XP_021118303.1 | 5 - 6 | 716 | 73 | ||
| XM_021262645.1 | XP_021118304.1 | 4 - 5 | 390 | 73 | ||
| XM_021262647.1 | XP_021118306.1 | 5 - 6 | 719 | 73 | ||

Virma
Genomic Map