Assay Details
| Gene Symbol: Wasf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850716.3 | XP_004850773.1 | 3 - 4 | 500 | 66 | |
| XM_021263149.1 | XP_021118808.1 | 3 - 4 | 327 | 66 | ||
More Information
Set Membership: |
|

Wasf2
Genomic Map