Assay Details
| Gene Symbol: Gjb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850911.3 | XP_004850968.1 | 1 - 2 | 330 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004850911.3 transcripts.
Set Membership: |
|

Gjb3
Genomic Map