Assay Details
| Gene Symbol: LOC101712794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858465.3 | XP_004858522.3 | 2 - 3 | 285 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101712794| Gene Symbol: LOC101712794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858465.3 | XP_004858522.3 | 2 - 3 | 285 | 65 | |
Set Membership: |
|