Assay Details
| Gene Symbol: Thap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858970.3 | XP_004859027.1 | 1 - 2 | 286 | 59 | |
| XM_013077447.2 | XP_012932901.1 | 1 - 2 | 286 | 59 | ||
More Information
Set Membership: |
|

Thap8
Genomic Map