Assay Details
| Gene Symbol: Thap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858970.3 | XP_004859027.1 | 2 - 3 | 482 | 83 | |
| XM_013077447.2 | XP_012932901.1 | 2 - 3 | 482 | 83 | ||
| XM_021239537.1 | XP_021095196.1 | 2 - 3 | 498 | 83 | ||
| XM_021239538.1 | XP_021095197.1 | 2 - 3 | 468 | 83 | ||
More Information
Set Membership: |
|

Thap8
Genomic Map