Assay Details
| Gene Symbol: Thap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858970.3 | XP_004859027.1 | 3 - 4 | 877 | 86 | |
| XM_021239537.1 | XP_021095196.1 | 3 - 4 | 893 | 86 | ||
| XM_021239538.1 | XP_021095197.1 | 3 - 4 | 863 | 86 | ||
More Information
Set Membership: |
|

Thap8
Genomic Map