Assay Details
| Gene Symbol: LOC101700851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861170.2 | XP_004861227.1 | 7 - 8 | 1265 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101700851| Gene Symbol: LOC101700851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861170.2 | XP_004861227.1 | 7 - 8 | 1265 | 73 | |
Set Membership: |
|