Assay Details
| Gene Symbol: LOC101701927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862992.3 | XP_004863049.1 | 2 - 3 | 275 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004862992.3 transcripts.
Set Membership: |
|

LOC101701927
Genomic Map