Assay Details
| Gene Symbol: LOC101700069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871369.2 | XP_004871426.1 | 1 - 2 | 1752 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004871369.2 transcripts.
Set Membership: |
|

LOC101700069
Genomic Map