Assay Details
| Gene Symbol: LOC101700069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871369.2 | XP_004871426.1 | 3 - 4 | 1922 | 100 | |
| XM_013068026.2 | XP_012923480.1 | 9 - 10 | 1155 | 100 | ||
| XM_021249482.1 | XP_021105141.1 | 8 - 9 | 1056 | 100 | ||
More Information
Set Membership: |
|

LOC101700069
Genomic Map