Assay Details
| Gene Symbol: Cers1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873397.3 | XP_004873454.1 | 6 - 7 | 1029 | 79 | |
| XM_021250988.1 | XP_021106647.1 | 6 - 7 | 846 | 79 | ||
More Information
Set Membership: |
|

Cers1
Genomic Map