Assay Details
| Gene Symbol: Ntf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066311.2 | XP_012921765.1 | 2 - 3 | 215 | 114 | |
| XM_021245962.1 | XP_021101621.1 | 2 - 3 | 503 | 114 | ||
| XM_021245963.1 | XP_021101622.1 | 2 - 3 | 200 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013066311.2, XM_021245962.1, XM_021245963.1 transcripts.
Set Membership: |
|

Ntf4
Genomic Map