Assay Details
| Gene Symbol: Alg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071999.2 | XP_012927453.1 | 3 - 4 | 1409 | 60 | |
| XM_021257811.1 | XP_021113470.1 | 3 - 4 | 1448 | 60 | ||
More Information
Set Membership: |
|

Alg11
Genomic Map