Assay Details
| Gene Symbol: LOC101710435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073652.2 | XP_012929106.1 | 39 - 40 | 5011 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101710435| Gene Symbol: LOC101710435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073652.2 | XP_012929106.1 | 39 - 40 | 5011 | 95 | |
Set Membership: |
|