Assay Details
| Gene Symbol: Znf280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077889.2 | XP_012933343.1 | 7 - 8 | 857 | 68 | |
| XM_021240554.1 | XP_021096213.1 | 7 - 8 | 684 | 68 | ||
| XM_021240555.1 | XP_021096214.1 | 7 - 8 | 858 | 68 | ||
More Information
Set Membership: |
|

Znf280c
Genomic Map