Assay Details
| Gene Symbol: LOC101699453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247515.1 | XP_021103174.1 | 1 - 2 | 1301 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101699453| Gene Symbol: LOC101699453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247515.1 | XP_021103174.1 | 1 - 2 | 1301 | 86 | |
Set Membership: |
|