Assay Details
| Gene Symbol: CUNH17orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248913.1 | XP_021104572.1 | 1 - 2 | 52 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021248913.1 transcripts.
Set Membership: |
|

CUNH17orf50
Genomic Map