Assay Details
| Gene Symbol: Galnt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837329.3 | XP_004837386.1 | 6 - 7 | 777 | 55 | |
| XM_013067539.2 | XP_012922993.1 | 5 - 6 | 464 | 55 | ||
More Information
Set Membership: |
|

Galnt16
Genomic Map