Assay Details
Gene Symbol: CUNH9orf85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838132.3 | XP_004838189.1 | 2 - 3 | 319 | 94 | |
XM_013069676.2 | XP_012925130.1 | 2 - 3 | 515 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
